1-(Pyrimidin-2-yl)piperidin-4-ol 95%

Supplier: Ambeed
Warning

A206884-10G A206884-1G A206884-5G A206884-250MG
103817-742EA 41 USD
103817-742 103848-422 76785-530 77086-686 77086-688 77667-734
1-(Pyrimidin-2-yl)piperidin-4-ol 95%
1-(Pyrimidin-2-yl)piperidin-4-ol

Precaución: for Research Use Only

Formula: C₉H₁₃N₃O
MW: 179.2200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864742
CAS Number: 893755-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR