1-(Pyrimidin-2-yl)ethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A153995-100MG A153995-250MG A153995-1G
103804-592EA 334 USD
103804-592 103842-772 77162-904 77162-906 77300-038
1-(Pyrimidin-2-yl)ethanamine hydrochloride 97%
1-(Pyrimidin-2-yl)ethanamine hydrochloride

Precaución: Inert atmosphere, room temperature

Formula: C₆H₁₀ClN₃
MW: 159.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27922039
CAS Number: 1616809-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR