1-(4-Pyridyl)piperazine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-(1-Piperazinyl)pyridine, 1-(Pyridin-4-yl)piperazine
A555975-1G
A555975-250MG
A555975-100G
A555975-10G
103828-714EA
6
USD
103828-714
103839-850
76843-942
77017-714
77684-400
77684-401
1-(4-Pyridyl)piperazine 97%
1-(4-Pyridyl)piperazine
Precaución: For research use only.
Formula:
C₉H₁₃N₃ MW: 163.22 g/mol Melting Pt: 137…141 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00040745 CAS Number: 1008-91-9 UN: 3259 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...