1-(Pyridin-4-yl)ethanamine dihydrochloride 97%

Supplier: Ambeed
Warning

A106711-10G A106711-250MG A106711-1G
103797-060EA 35 USD
103797-060 103822-816 76982-464 76982-466 77283-498
1-(Pyridin-4-yl)ethanamine dihydrochloride 97%
1-(Pyridin-4-yl)ethanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂Cl₂N₂
MW: 195.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09028144
CAS Number: 174132-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR