1-(Pyridin-4-yl)cyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

A120337-100MG A120337-250MG A120337-1G A120337-5G
103799-918EA 199 USD
103799-918 76822-366 76822-368 77312-026 77312-028
1-(Pyridin-4-yl)cyclopropanecarboxylic acid 97%
1-(Pyridin-4-yl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864787
CAS Number: 920297-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR