1-(Pyridin-3-yl)-1H-pyrazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A100879-250MG A100879-1G A100879-5G
103795-670EA 52 USD
103795-670 103821-362 76982-168 77296-878 77296-880
1-(Pyridin-3-yl)-1H-pyrazole-4-carboxylic acid 98%
1-(Pyridin-3-yl)-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇N₃O₂
Boiling Pt: 417.7±25.0 °C (760 mm Hg)
Flash Pt: 206.4 °C
Storage Temperature: Ambient
MDL Number: MFCD18260889
CAS Number: 1014631-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR