1-(Pyridin-2-yl)propan-2-one 98% mix

Supplier: Ambeed
Warning

A109021-100MG A109021-10G A109021-250MG A109021-1G A109021-5G
103797-430EA 14 USD
103797-430 103823-218 77233-402 77233-404 77233-406 77283-798 77310-048
1-(Pyridin-2-yl)propan-2-one 98% mix
1-(Pyridin-2-yl)propan-2-one

Precaución: for Research Use Only

Formula: C₈H₉NO
MW: 135.1632 g/mol
Flash Pt: 85.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00092717
CAS Number: 6302-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR