1-(Pyridin-2-yl)piperazin-2-one 97%

Supplier: Ambeed
Warning

A174042-100MG A174042-250MG A174042-1G
103806-906EA 524 USD
103806-906 76974-164 76974-166 77082-488
1-(Pyridin-2-yl)piperazin-2-one 97%
1-(Pyridin-2-yl)piperazin-2-one

Precaución: for Research Use Only

Formula: C₉H₁₁N₃O
Boiling Pt: 432.5 °C (760 mm Hg)
Flash Pt: 215.4 °C
Storage Temperature: Ambient
MDL Number: MFCD07373443
CAS Number: 345310-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR