2-Acetylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Pyridin-2-yl)ethanone, Methyl 2-pyridyl ketone

A302917-25G A302917-1000G A302917-100G A302917-10G A302917-500G
103835-514EA 7 USD
103835-514 76838-204 77154-220 77154-222 77154-224 77154-226
2-Acetylpyridine 98%
2-Acetylpyridine

Precaución: for Research Use Only

Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 188…189 °C (760 mm Hg)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00006303
CAS Number: 1122-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR