2-(1-Hydroxyethyl)pyridine 98%

Supplier: Ambeed
Warning

A109607-10G A109607-25G A109607-1G A109607-5G
103797-532EA 10 USD
103797-532 103823-332 77233-482 77233-484 77310-142 77310-144
2-(1-Hydroxyethyl)pyridine 98%
2-(1-Hydroxyethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
Boiling Pt: 210.6 °C (760 mm Hg)
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD06245419
CAS Number: 18728-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR