1-(Pyridazin-4-yl)ethanone 98%

Supplier: Ambeed
Warning

A388934-250MG A388934-1G
103826-842EA 451 USD
103826-842 103837-882 76788-830 76841-244
1-(Pyridazin-4-yl)ethanone 98%
1-(Pyridazin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₆N₂O
Boiling Pt: 285.8 °C (760 mm Hg)
Melting Pt: 69…70 °C
Storage Temperature: Freezer
MDL Number: MFCD13193354
CAS Number: 50901-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR