1-(4-Methylphenyl)cyclopropanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-(p-Tolyl)cyclopropanecarboxylic acid

A141628-10G A141628-250MG A141628-1G A141628-5G
103803-030EA 36 USD
103803-030 77190-464 77190-466 77299-230 77299-232
1-(4-Methylphenyl)cyclopropanecarboxylic acid 98%
1-(4-Methylphenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.2100 g/mol
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00066918
CAS Number: 83846-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR