1-(Phenylsulfonyl)indole 98%

Supplier: Ambeed
Warning

A310819-100G A310819-10G A310819-1G A310819-500G A310819-25G A310819-5G
103835-822EA 6 USD
103835-822 77141-676 77141-678 77141-680 77141-682 77306-036 77306-038
1-(Phenylsulfonyl)indole 98%
1-(Phenylsulfonyl)indole

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₂S
Boiling Pt: 451.6 °C (760 mm Hg)
Melting Pt: 78…80 °C
Flash Pt: 226.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00134318
CAS Number: 40899-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR