1-(Oxetan-3-yl)piperazine 95%

Supplier: Ambeed
Warning

A124587-10G A124587-250MG A124587-1G A124587-5G A124587-25G
103800-554EA 11 USD
103800-554 76983-410 76983-412 77298-006 77298-008 77571-474
1-(Oxetan-3-yl)piperazine 95%
1-(Oxetan-3-yl)piperazine

Precaución: for Research Use Only

Formula: C₇H₁₄N₂O
Boiling Pt: 227.2 °C (760 mm Hg)
Flash Pt: 91.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD21847918
CAS Number: 1254115-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR