3'-Methylacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: Methyl m-tolyl ketone, 1-(m-Tolyl)ethanone, m-Methylacetophenone, 3′-Methylacetophenone

A182131-5G A182131-100G A182131-10G A182131-500G
103846-056EA 19 USD
103846-056 77083-590 77593-164 77593-166 77685-424
3'-Methylacetophenone 98%
3'-Methylacetophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 218…220 °C (760 mm Hg)
Melting Pt: 9 °C
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00008742
CAS Number: 585-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR