Cyclopropane-1,1-dicarboxylic acid mono methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Methoxycarbonyl)cyclopropanecarboxylic acid

A221325-100G A221325-10G A221325-1G A221325-25G A221325-5G A221325-500G
103818-914EA 9 USD
103818-914 103849-796 76691-826 76691-828 77303-588 77303-590 77303-592 77626-114
Cyclopropane-1,1-dicarboxylic acid mono methyl ester 97%
Cyclopropane-1,1-dicarboxylic acid mono methyl ester

Precaución: for Research Use Only

Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 245.3 °C (760 mm Hg)
Melting Pt: 47…51 °C
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD06800496
CAS Number: 113020-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR