1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Mesitylsulphonyl)-3-nitro-1H-1,2,4-triazole

A204347-10G A204347-25G A204347-1G A204347-5G
103817-546EA 11 USD
103817-546 103848-206 76800-466 76800-468 77086-368 77086-370
1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole 97%
1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₄O₄S
MW: 296.301 g/mol
Melting Pt: 134…139 °C
Flash Pt: 276.4 °C
Storage Temperature: Freezer
MDL Number: MFCD00009754
CAS Number: 74257-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR