1-(Isoxazol-5-yl)ethanone 95%

Supplier: Ambeed
Warning

A125446-100MG A125446-250MG A125446-1G
103800-678EA 207 USD
103800-678 76810-862 76810-864 77312-824
1-(Isoxazol-5-yl)ethanone 95%
1-(Isoxazol-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₅H₅NO₂
MW: 111.1 g/mol
Boiling Pt: 205.5 °C (760 mm Hg)
Flash Pt: 78.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16619203
CAS Number: 88511-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR