1-Acetylindoline 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Indolin-1-yl)ethanone

A279487-25G A279487-5G
103853-412EA 25 USD
103853-412 77093-346 77093-348
1-Acetylindoline 97%
1-Acetylindoline

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
MW: 161.2
Boiling Pt: 355.1 °C (760 mm Hg)
Melting Pt: 102…104 °C
Flash Pt: 174.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00022908
CAS Number: 16078-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR