1-(Hydroxymethyl)cyclopropanecarbonitrile 97%
Supplier: Ambeed
A497448-1G
A497448-100MG
A497448-250MG
A497448-5G
103828-266EA
31
USD
103828-266
103839-344
76843-314
77034-460
77034-462
77307-602
1-(Hydroxymethyl)cyclopropanecarbonitrile 97%
1-(Hydroxymethyl)cyclopropanecarbonitrile
Precaución: for Research Use Only
Formula:
C₅H₇NO |
MDL Number:
MFCD11042632 CAS Number: 98730-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...