1-(Difluoromethyl)-4-nitrobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 4-Nitrobenzal fluoride

A158247-25G A158247-1G A158247-5G
103805-110EA 17 USD
103805-110 103843-318 76783-704 77300-320 77319-172
1-(Difluoromethyl)-4-nitrobenzene 95%
1-(Difluoromethyl)-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
Boiling Pt: 240.7 °C (760 mm Hg)
Melting Pt: 33…35 °C
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16659620
CAS Number: 29848-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR