1-Cyclopropylmethyl-1H-pyrazole-4-boronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Cyclopropylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

A200378-100MG A200378-250MG
103817-194EA 103 USD
103817-194 103847-820 76975-228 76975-230
1-Cyclopropylmethyl-1H-pyrazole-4-boronic acid, pinacol ester 97%
1-Cyclopropylmethyl-1H-pyrazole-4-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₁BN₂O₂
Storage Temperature: Freezer
MDL Number: MFCD15474887
CAS Number: 1000801-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR