1-(Cyclopropylcarbonyl)piperazine 97%

Supplier: Ambeed
Danger

A325141-1G A325141-25G A325141-5G A325141-100G A325141-10G
103825-390EA 6 USD
103825-390 103836-334 76839-260 76839-262 76839-264 77114-772 77114-774
1-(Cyclopropylcarbonyl)piperazine 97%
1-(Cyclopropylcarbonyl)piperazine

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O
Boiling Pt: 305.5 °C (760 mm Hg)
Flash Pt: 138.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06369645
CAS Number: 59878-57-8
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR