3-Methoxybenzyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 1-(Chloromethyl)-3-methoxybenzene, 3-(Chloromethyl)anisole

A202463-1G A202463-250MG
103848-034EA 8 USD
103848-034 77112-204 77611-960
3-Methoxybenzyl chloride 97%
3-Methoxybenzyl chloride

Precaución: for Research Use Only

Formula: C₈H₉ClO
Boiling Pt: 124 °C (760 mm Hg)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000907
CAS Number: 824-98-6
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR