4,5-Dimethoxy-2-nitrobenzyl bromide 96%

Supplier: Ambeed
Danger

A273941-1G A273941-10G
103813-168EA 45 USD
103813-168 103853-094 77092-910 77289-472
4,5-Dimethoxy-2-nitrobenzyl bromide 96%
4,5-Dimethoxy-2-nitrobenzyl bromide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₄
Boiling Pt: 372.3 °C (760 mm Hg)
Melting Pt: 131…133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192064
CAS Number: 53413-67-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR