α-Bromo-4-(difluoromethoxy)toluene 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-4-(difluoromethoxy)benzene, p-(Difluoromethoxy)benzyl bromide, 4-(Difluoromethoxy)benzyl bromide

A189044-25G A189044-5G A189044-10G A189044-250MG
103816-150EA 26 USD
103816-150 103846-744 77084-492 77084-494 77097-080 77097-082
α-Bromo-4-(difluoromethoxy)toluene 98%
α-Bromo-4-(difluoromethoxy)toluene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
MW: 237.04 g/mol
Boiling Pt: 237.9 °C (760 mm Hg)
Flash Pt: 117.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00085007
CAS Number: 3447-53-8
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR