α-Bromo-3-iodotoulene 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-3-iodobenzene, α-Bromo-m-iodotoulene, 3-Iodobenzyl bromide

A547856-25G A547856-5G A547856-10G A547856-250MG A547856-1G
103828-674EA 20 USD
103828-674 103839-796 76843-862 76843-864 77103-482 77103-484 77307-790
α-Bromo-3-iodotoulene 98%
α-Bromo-3-iodotoulene

Precaución: for Research Use Only

Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 281.2 °C (760 mm Hg)
Melting Pt: 46…51 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00019016
CAS Number: 49617-83-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR