3,5-Di-tert-butylbenzyl bromide 98%

Supplier: Ambeed
Danger

A507058-5G A507058-100MG A507058-10G A507058-1G A507058-250MG
103828-340EA 15 USD
103828-340 103839-444 76843-426 77279-732 77279-734 77307-642 77617-166
3,5-Di-tert-butylbenzyl bromide 98%
3,5-Di-tert-butylbenzyl bromide

Precaución: for Research Use Only

Formula: C₁₅H₂₃Br
Melting Pt: 29…34 °C
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD03701623
CAS Number: 62938-08-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR