3,5-Dimethoxybenzyl bromide 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-(Bromomethyl)-3,5-dimethoxybenzene
A586795-1G
A586795-100G
A586795-10G
A586795-250MG
A586795-25G
103828-900EA
11
USD
103828-900
103840-046
76844-192
77117-212
77117-214
77117-216
77307-886
3,5-Dimethoxybenzyl bromide 97%
3,5-Dimethoxybenzyl bromide
Precaución: for Research Use Only
Formula:
C₉H₁₁BrO₂ MW: 231.0864 g/mol Melting Pt: 69…70 °C Flash Pt: 129.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01321368 CAS Number: 877-88-3 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...