1-(Bromomethyl)-3,5-dichlorobenzene 95%

Supplier: Ambeed
Danger

A107534-100MG A107534-250MG A107534-25G A107534-1G A107534-5G
103797-206EA 41 USD
103797-206 103822-974 77270-662 77270-664 77270-666 77283-628 77309-832
1-(Bromomethyl)-3,5-dichlorobenzene 95%
1-(Bromomethyl)-3,5-dichlorobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MW: 239.92 g/mol
Boiling Pt: 260.852 °C (760 mm Hg)
Flash Pt: 126.228 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10698783
CAS Number: 7778-01-0
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR