3-(Methylsulfonyl)benzyl bromide 96%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-3-(methylsulphonyl)benzene, 3-(Bromomethyl)phenyl methyl sulphone

A144030-100MG A144030-250MG A144030-1G A144030-5G
103803-358EA 228 USD
103803-358 103841-434 76811-890 76811-892 77316-672 77316-674
3-(Methylsulfonyl)benzyl bromide 96%
3-(Methylsulfonyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₈H₉BrO₂S
MW: 249.125 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12025160
CAS Number: 82657-76-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR