α-Bromo-2-iodotoulene 98%

Supplier: Ambeed
Danger

Synonyms: α-Bromo-o-iodotoulene, 2-Iodobenzyl bromide, 1-(Bromomethyl)-2-iodobenzene

A675303-5G A675303-10G A675303-1G A675303-25G
103840-520EA 19 USD
103840-520 76844-720 77104-746 77308-108 77308-110
α-Bromo-2-iodotoulene 98%
α-Bromo-2-iodotoulene

Precaución: for Research Use Only

Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 278.8 °C (760 mm Hg)
Melting Pt: 54…60 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236046
CAS Number: 40400-13-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR