2-Chloro-3-fluorobenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-2-chloro-3-fluorobenzene

A119620-100G A119620-10G A119620-250MG A119620-25G A119620-1G A119620-5G
103799-802EA 12 USD
103799-802 77108-490 77108-492 77108-494 77108-496 77311-912 77311-914
2-Chloro-3-fluorobenzyl bromide 98%
2-Chloro-3-fluorobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₅BrClF
Boiling Pt: 227.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09038462
CAS Number: 874285-19-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR