2,4-Dichlorobenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-2,4-dichlorobenzene

A231289-1G A231289-25G A231289-5G A231289-10G
103810-770EA 14 USD
103810-770 103850-518 77089-402 77089-404 77089-406 77249-578
2,4-Dichlorobenzyl bromide 98%
2,4-Dichlorobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MW: 239.9246 g/mol
Flash Pt: 126.228 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03208523
CAS Number: 20443-99-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR