1-(Bromomethyl)-2,3-dimethylbenzene 95%

Supplier: Ambeed
Danger

Synonyms: 2,3-Dimethylbenzyl bromide

A172038-10G A172038-250MG A172038-25G A172038-5G
103806-688EA 13 USD
103806-688 103844-970 76690-212 76690-214 77301-178 77301-180
1-(Bromomethyl)-2,3-dimethylbenzene 95%
1-(Bromomethyl)-2,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁Br
MW: 199.0876 g/mol
Boiling Pt: 240 °C (760 mm Hg)
Flash Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03844741
CAS Number: 81093-21-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR