1-(Boc-amino)cyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Boc-Amino)cyclopropanecarboxylic acid, 1-Aminocyclopropane-1-carboxylic acid, N-BOC protected, 1-[(tert-Butoxycarbonyl)amino]cyclopropanecarboxylic acid

A251472-25G A251472-5G A251472-10G A251472-1G A251472-100G A251472-500G
103811-984EA 7 USD
103811-984 103851-826 77091-210 77091-212 77249-962 77304-558 77746-397 77746-398
1-(Boc-amino)cyclopropanecarboxylic acid 97%
1-(Boc-amino)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
Boiling Pt: 347.6 °C (760 mm Hg)
Melting Pt: 178 °C
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00083257
CAS Number: 88950-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR