But-3-en-1-amine, N-BOC protected 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Boc-amino)-3-butene

A326774-1G A326774-100MG A326774-10G A326774-250MG A326774-5G
103825-446EA 27 USD
103825-446 103836-398 76839-366 76875-946 76875-948 76875-950 77306-352
But-3-en-1-amine, N-BOC protected 97%
But-3-en-1-amine, N-BOC protected

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MW: 171.237 g/mol
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11975862
CAS Number: 156731-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR