1-(Benzyloxy)-6-oxo-1,6-dihydropyridine-2-carboxylic acid 96%
Supplier: Ambeed
|
Warning
|
A170986-250MG
A170986-1G
103806-570EA
43
USD
103806-570
103844-844
76872-434
77081-970
1-(Benzyloxy)-6-oxo-1,6-dihydropyridine-2-carboxylic acid 96%
1-(Benzyloxy)-6-oxo-1,6-dihydropyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₁NO₄ MW: 245.23 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20270106 CAS Number: 210366-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...