Fmoc-NH-PEG3-CH2COOH 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13-tetraoxa-4-azapentadecan-15-oic acid
A117639-100MG
A117639-10G
A117639-250MG
A117639-25G
A117639-1G
A117639-5G
103798-914EA
55
USD
103798-914
103824-802
77161-112
77161-114
77161-116
77161-118
77311-586
77311-588
Fmoc-NH-PEG3-CH2COOH 95%
Fmoc-NH-PEG3-CH2COOH
Precaución: for Research Use Only
Formula:
C₂₃H₂₇NO₇ Melting Pt: 46 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD13182400 CAS Number: 139338-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...