9-Acetylcarbazole 99%

Supplier: Ambeed
Warning

A264439-1G A264439-5G A264439-250MG
103812-678EA 9 USD
103812-678 103852-570 77092-236 77092-238 77113-764
9-Acetylcarbazole 99%
9-Acetylcarbazole

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO
Boiling Pt: 315.5 °C (760 mm Hg)
Melting Pt: 77…79 °C
Flash Pt: 144.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00087027
CAS Number: 574-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR