1-(6-Nitropyridin-3-yl)piperazine 95%
Supplier: Ambeed
|
Warning
|
A100484-250MG
A100484-1G
103795-470EA
23
USD
103795-470
103799-216
76821-208
77296-716
1-(6-Nitropyridin-3-yl)piperazine 95%
1-(6-Nitropyridin-3-yl)piperazine
Precaución: for Research Use Only
Formula:
C₉H₁₂N₄O₂ Boiling Pt: 422.8 °C (760 mm Hg) Flash Pt: 209.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09041112 CAS Number: 775288-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...