1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol 97%

Supplier: Ambeed
Warning

Synonyms: alpha-Methyl-6-nitropiperonyl alcohol, 1-(6-Nitrobenzo[d][1,3]dioxol-5-yl)ethanol, alpha-Methyl-6-nitro-1,3-benzodioxole-5-methanol

A171977-100MG A171977-250MG A171977-1G
103806-684EA 64 USD
103806-684 103844-966 77273-748 77273-750 77301-176
1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol 97%
1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MW: 211.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08274663
CAS Number: 159873-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR