1-(6-Methylpyridin-3-yl)hydrazine hydrochloride 95%

Supplier: Ambeed
Warning

A199461-100MG A199461-250MG A199461-1G
103817-106EA 66 USD
103817-106 103847-730 76814-224 76814-226 77085-772
1-(6-Methylpyridin-3-yl)hydrazine hydrochloride 95%
1-(6-Methylpyridin-3-yl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₀ClN₃
MW: 159.6200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10698812
CAS Number: 896133-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR