1-(6-Methoxynaphthalen-2-yl)propan-1-one 97%

Supplier: Ambeed
Warning

A362117-5G A362117-100G A362117-500G
103826-302EA 6 USD
103826-302 103837-316 76840-530 77003-370 77003-372
1-(6-Methoxynaphthalen-2-yl)propan-1-one 97%
1-(6-Methoxynaphthalen-2-yl)propan-1-one

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Boiling Pt: 343.4 °C (760 mm Hg)
Melting Pt: 110…112 °C
Flash Pt: 158.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00021645
CAS Number: 2700-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR