1-(6-Chloropyridin-2-yl)ethanone 97%

Supplier: Ambeed
Warning

A363205-1G A363205-5G A363205-250MG
103826-336EA 72 USD
103826-336 103837-356 76840-576 76840-578 76989-818
1-(6-Chloropyridin-2-yl)ethanone 97%
1-(6-Chloropyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MW: 155.58 g/mol
Boiling Pt: 249.931 °C (760 mm Hg)
Flash Pt: 104.955 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07375070
CAS Number: 152356-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR