1-(6-Bromo-2-pyridinyl)-1-ethanol 95%

Supplier: Ambeed
Warning

Synonyms: 1-(6-Bromopyridin-2-yl)ethanol

A158661-100MG A158661-10G A158661-250MG A158661-1G A158661-5G
103805-146EA 156 USD
103805-146 103843-362 76997-672 76997-674 76997-676 77300-340 77300-342
1-(6-Bromo-2-pyridinyl)-1-ethanol 95%
1-(6-Bromo-2-pyridinyl)-1-ethanol

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12198456
CAS Number: 139163-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR