1-(6-(Trifluoromethyl)pyridin-3-yl)ethanone 97%

Supplier: Ambeed
Warning

Synonyms: 5-Acetyl-2-(trifluoromethyl)pyridine

A161402-100MG A161402-10G A161402-250MG A161402-1G A161402-25G A161402-5G
103805-482EA 45 USD
103805-482 103843-720 76812-756 76812-758 76812-760 77319-692 77319-694 77319-696
1-(6-(Trifluoromethyl)pyridin-3-yl)ethanone 97%
1-(6-(Trifluoromethyl)pyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆F₃NO
MW: 189.1346 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375085
CAS Number: 358780-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR