1-(5-Methyl-1H-pyrrol-3-yl)ethanone 97%
Supplier: Ambeed
|
Warning
|
A164741-100MG
A164741-250MG
A164741-1G
103805-898EA
254
USD
103805-898
77191-212
77191-214
77300-732
1-(5-Methyl-1H-pyrrol-3-yl)ethanone 97%
1-(5-Methyl-1H-pyrrol-3-yl)ethanone
Precaución: Sealed in dry, at room temperature.
Formula:
C₇H₉NO MW: 123.152 g/mol Boiling Pt: 249.3±20.0 °C (760 mm Hg) Melting Pt: 113 °C Flash Pt: 111 °C Storage Temperature: Ambient |
MDL Number:
MFCD19219386 CAS Number: 6115-72-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...