1-(5-hydroxypyridin-2-yl)ethanone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Hydroxypyridin-2-yl)ethanone

A236498-100MG A236498-250MG A236498-1G A236498-5G
103811-100EA 55 USD
103811-100 103850-872 76786-410 76786-412 77089-918 77089-920
1-(5-hydroxypyridin-2-yl)ethanone 97%
1-(5-hydroxypyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₇NO₂
Boiling Pt: 383.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD10697710
CAS Number: 67310-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR