1-(5-Hydroxypyrazin-2-yl)ethanone 98%

Supplier: Ambeed
Warning

A100360-100MG A100360-1G
103795-384EA 336 USD
103795-384 103821-084 76982-126 77282-480
1-(5-Hydroxypyrazin-2-yl)ethanone 98%
1-(5-Hydroxypyrazin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MW: 138.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12400925
CAS Number: 1159813-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR